Learn More
AdipoGen D-chiro-Inositol (1 g)
SDP
Supplier:  AdipoGen CDXD0798G001
Encompass_Preferred
Chemical. CAS: 643-12-9. Formula: C6H12O6. MW: 180.16. D-chiro Inositol is a insulin sensitizing agent. It is a secondary messenger in the signal transduction of insulin. D-chiro Inositol reduces the effects of polycystic ovary syndrome (PCOS) activity in cells. D-chiro Inositol is a mediator of insulin action via accelerating the dephosphorylation of glycogen synthase and pyruvate dehydrogenase and rate limiting enzymes of non-oxidative and oxidative glucose disposal. As a result of these actions D-chiro Inositol functions to reduce insulin resistance and increase sensitivity and glucose disposal. Can also be used as a building block for organic synthesis.
Catalog No. 50-205-4401
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.