Learn More
Abcam D-Cycloserine, NMDA Glycine Site Agonist, 99% Purity, 5 g
Description
- Molecular weight: 102.09 Da
- Purity: >99%
- Chemical structure: (R)-4-amino-1,2-oxazolidin-3-one (100%)
- Activity: Partial agonist at the NMDA receptor glycine recognition site
- Biological effect: Enhances learning and memory in vivo; performance enhancer in cognitive models
- CAS Number: 68-41-7
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | NMDA Receptor (Glycine Recognition Site) |
| Substrate Properties | Chemical Substrate |
| Alternate Part Number | ab146677 |
| Buffer | 100% (R)-4-amino-1,2-oxazolidin-3-one |
| Content And Storage | Store at -20°C. |
| Form | Powder |
| Formulation | 100% (R)-4-amino-1,2-oxazolidin-3-one |
| Quantity | 5 g |
| For Use With (Application) | NMDA receptor agonist, learning and memory research, cognitive models |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.