Learn More
AdipoGen d-Desthiobiotin
SDP
Supplier:  AdipoGen CDXD0272M500
Encompass_Preferred
Chemical. CAS 533-48-2. Formula C10H18N2O3. Molecular Weight 214.26. D-Desthiobiotin is a protein cross-linking agent and a precursor in the synthesis of biotin. Desthiobiotin is a modified form of biotin that binds less tightly to avidin and streptavidin than biotin while still providing excellent specificity in affinity purification methods. Unlike biomolecules that are labeled with biotin, proteins and other targets that are labeled with desthiobiotin can be eluted without harsh, denaturing conditions. D-Desthiobiotin is used in affinity chromatography and protein chromatography, and can be used for protein and cell labeling, detection and isolation. It has been used in the preparation of agarose matrices for affinity-based isolation of streptavidin-fluorophore conjugates. Desthiobiotin is an efficient competitor for the mild elution of Strep-tag and TwinStrep-tag fusion proteins from Strep-Tactin columns. For elution 2.5 mM desthiobiotin is added to the buffer.
Catalog No. 50-256-1152
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.