Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ D-dimer Monoclonal Antibody (28)
GREENER_CHOICE

Catalog No. 01670107
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-107 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-107 Supplier Invitrogen™ Supplier No. ABS0152802
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABS 015-28-02 detects D-dimer in human and dog samples. ABS 015-28-02 has been successfully used in ELISA procedures. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

Fibrinogen is the main protein of blood coagulation system. It consists of two identical subunits that contain three polypeptide chains: alpha, beta and gamma. The process of blood coagulation results in the activation of fibrinogen into fibrin by thrombin and fibrin polymerization. Fibrin clot is then digested by plasmin, and fibrin degradation products of different molecular weights are released into the bloodstream.
TRUSTED_SUSTAINABILITY

Specifications

Antigen D-dimer
Applications ELISA, Western Blot
Classification Monoclonal
Clone 28
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene FGA
Gene Accession No. P02671, P68213
Gene Alias carboxy-terminal region; ENSMUSG00000059807; FGA; Fib; Fib2; fibrinogen A-alpha-chain; fibrinogen alpha chain; fibrinogen, A alpha polypeptide; Fibrinopeptide A; MGC119422; MGC119423; MGC119425
Gene Symbols FGA
Host Species Mouse
Immunogen Purified human D-dimer.
Purification Method Protein A/G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2243, 475473
Target Species Human, Canine
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.