Learn More
Enzo Life Sciences N,N-Dimethylsphingosine, D-erythro (5 mg). CAS: 119567-63-4
Supplier: Enzo Life Sciences BMLSL1050005
Potent and specific inhibitor of sphingosine kinase (IC50=5µM) which blocks conversion of sphingosine to sphingosine-1-phosphate (BML-SL140). Inhibitor of protein kinase C (PKC) which also stimulates Src-kinase. May affect expression of cell surface selectins, which in turn mediate leukocyte or tumor cell adhesion to endothelial cells and platelets. Induces apoptosis. Stereospecifically induces EGF receptor autophosphorylation producing EGF-like activity in vitro in the absence of EGF. Produced endogenously via ceramide catabolism. May be a target for neuropathic pain therapy. Alternative name: N,N-Dimethyl-D-erythro-sphingosine. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.