Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam D-erythro-Dihydrosphingosine-1-Phosphate, Receptor Ligand, Purity >98%, 5 mg

Catalog No. 502706444
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6444 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6444 Supplier Abcam Supplier No. AB1417505MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent sphingosine 1-phosphate receptor ligand with anti-fibrotic activity in fibroblasts.

  • Molecular Weight: 381.5 Da
  • Purity: >98%
  • Form: Solid
  • CAS Number: 19794-97-9
  • Activity: Sphingosine 1-phosphate (S1P) receptor ligand (Ki = 15 nM); acts as an anti-fibrotic in fibroblasts and may be used as a negative control for intracellular effects of S1P (ab141748)
  • Composition: 100% [(2S,3R)-2-amino-3-hydroxyoctadecyl] dihydrogen phosphate

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% [(2S,3R)-2-amino-3-hydroxyoctadecyl] dihydrogen phosphate
Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule Sphingosine 1-phosphate receptor
Formulation 100% [(2S,3R)-2-amino-3-hydroxyoctadecyl] dihydrogen phosphate
Quantity 5 mg
For Use With (Application) Sphingosine 1-phosphate receptor ligand, negative control for intracellular effects of S1P
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.