Learn More
Abcam D-erythro-Dihydrosphingosine-1-Phosphate, Receptor Ligand, Solid, 1 mg
Description
- Molecular Weight: 381.5 Da
- Purity: >98%
- Form: Solid
- Activity: Sphingosine 1-phosphate (S1P) receptor ligand (Ki = 15 nM)
- Application: May be used as negative control for intracellular effects of S1P (ab141748); anti-fibrotic effect in fibroblasts
- Composition: 100% [(2S,3R)-2-amino-3-hydroxyoctadecyl] dihydrogen phosphate
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Sphingosine 1-Phosphate Receptor |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Buffer | 100% [(2S,3R)-2-amino-3-hydroxyoctadecyl] dihydrogen phosphate |
| Formulation | 100% [(2S,3R)-2-amino-3-hydroxyoctadecyl] dihydrogen phosphate |
| Quantity | 1 mg |
| For Use With (Application) | Sphingosine 1-phosphate receptor ligand, negative control for intracellular effects of S1P |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.