Learn More
Leica Biosystems D-FORMALIZER PAD 8X10 25/PK
Supplier:  Leica Biosystems 38007530810
Encompass_Preferred
D-Formalizer Pads reduce formaldehyde exposure by absorbing and neutralizing any excess formalin. The durable blue surface of the pads provides an excellent contrast to even small pieces of tissue. The Pads are also useful for soaking up leakage and de-odourising vapours in other applications. They can be used to line shelves which hold specimen containers that may seep formaldehyde around the lid, or to line transport boxes where leaks may occur. D-Formalizer Pads are available in various sizes and also in a convenient roll so that lengths can be cut as required.
Catalog No. NC9025368
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.