Learn More
Abcam D-Fructose 1,6-Diphosphate Trisodium Salt, >98% Purity, 1 g
Description
- Molecular weight: 406.06 g/mol
- Purity: >98%
- General description: Common metabolic sugar. Precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway.
- Biological activity: Allosteric activator of enzymes such as pyruvate kinase and NAD+-dependent L-(+)-lactate dehydrogenase. Inhibitor of acetate kinase. Used as a substrate to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolase(s) and fructose-1,6-bisphosphatase(s).
- Composition: 100% D-Fructose 1,6-diphosphate trisodium salt
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | D-Fructose 1,6-diphosphate |
| Substrate | Fructose-1,6-bisphosphate |
| Content And Storage | Store at -20°C. |
| Form | Powder |
| Shipping Condition | Ambient |
| Quantity | 1 g |
| For Use With (Application) | Enzyme characterization, glycolytic pathway research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.