Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam D-Fructose 1,6-Diphosphate Trisodium Salt, >98% Purity, 5 g

Catalog No. 502705342
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5342 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5342 Supplier Abcam Supplier No. AB1452765G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Common metabolic sugar with >98% purity, used as a key substrate and activator in metabolic pathways.

  • Molecular Weight: 406.06 g/mol
  • Purity: >98%
  • Description: Common metabolic sugar; precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway.
  • Activity: Allosteric activator of enzymes such as pyruvate kinase and NAD+-dependent L-(+)-lactate dehydrogenase; inhibitor of acetate kinase.
  • Application: Used as a substrate to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolase(s) and fructose-1,6-bisphosphatase(s).
  • Storage: Store at -20°C.
  • Shipping: Ambient - Can Ship with Ice.
  • Composition: 100% D-Fructose 1,6-diphosphate trisodium salt.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Fructose 1,6-bisphosphate
Content And Storage Store at -20°C.
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Substrate Fructose-1,6-bisphosphate
Formulation 100% D-Fructose 1,6-diphosphate trisodium salt
Quantity 5 g
For Use With (Application) Substrate for identifying and characterizing enzymes, metabolic pathway research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.