Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam D-Fructose 6-phosphate Disodium Salt Hydrate, >98% Purity, 500 mg

Catalog No. 502705207
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5207 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5207 Supplier Abcam Supplier No. AB145419500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Sugar intermediate of the glycolytic pathway with >98% purity.

  • Molecular weight: 304.1 g/mol
  • Purity: >98%
  • Description: Sugar intermediate of the glycolytic pathway produced by the isomerization of glucose-6-phosphate by phosphoglucoisomerase

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Content And Storage Store at -20°C.
Form Solid
Target Molecule D-Fructose 6-Phosphate
Shipping Condition Ambient
Quantity 500 mg
For Use With (Application) Glycolytic pathway research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.