Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-D-(-)-Fructose, MP Biomedicals™

Catalog No. MP210218601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210218601 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210218601 Supplier MP Biomedicals, Inc Supplier No. 0210218601
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Found in many plants. β-D-(-)-Fructose, MP Biomedicals (fruit sugar) is a dietary monosaccharide. This simple sugar is absorbed directly into the bloodstream during digestion. It is used for the diagnosis of cancer, as cancer cells show a higher level of

The three dietary monosaccharides (fructose, glucose and galactose) are absorbed directly into the bloodstream during digestion. Fructose is a 6-carbon polyhydroxyketone. It is an isomer of glucose which can be anaerobically fermented by yeast or bacteria.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
Synonym d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
PubChem CID 5984
ChEBI CHEBI:48095
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Melting Point 110°C to 120°C
Density 1.6g/mL (Lit.)
Quantity 1 kg
Solubility Information Soluble in water and alcohols.
Formula Weight 180.2
Percent Purity ≥99%
Physical Form Powder
Recommended Storage Room Temperature (15–30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.