accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)OH(R)OH(S)OHOOH

CAS RN 57-48-7

IUPAC Name: (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
Synonyms: keto-D-fructose
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: BJHIKXHVCXFQLS-UYFOZJQFSA-N
SMILES: OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO
Molecular Weight (g/mol) 
  • (25)
  • (18)
  • (1)
Grade 
  • (1)
  • (1)
  • (5)
  • (18)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (6)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (11)
  • (7)
Physical Form 
  • (2)
  • (4)
  • (11)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (21)
  • (11)
  • (1)
  • (2)
  • (3)
  • (17)
  • (11)
  • (2)
  • (10)
  • (1)
  • (10)
  • (1)
  • (25)
  • (18)
  • (1)
  • (1)
  • (1)
  • (5)
  • (18)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (11)
  • (7)
  • (2)
  • (4)
  • (11)
  • (3)
  • (1)