Learn More
Medchemexpress LLC D-galactopyranose, 2,3,4,6-tetrabenzoate | 627466-84-6 | MFCD09841311 | >96.0% | 596.58 g/mol | C34H28O10 | 250 MG
Supplier: Medchemexpress LLC HYW556877250MG
D-Galactopyranose,2,3,4,6-tetrabenzoate is a benzoyl-protected derivative of D-galactopyranose used as an intermediate and reagent in carbohydrate synthesis and glycobiology research. It is supplied as a solid laboratory reagent for protection/deprotection strategies and glycosylation workflows.
- Benzoyl-protected sugar derivative for protection strategies in carbohydrate synthesis.
- Synthetic intermediate suitable for glycosylation and derivatization reactions.
- Typically supplied as a solid powder for laboratory use.
- High purity (>96.0%) as reported by suppliers; verify certificate of analysis before use.
- Known identity for reference: CAS 627466-84-6 and molecular weight ≈596.58 g/mol.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.