Learn More
Medchemexpress LLC D-galactopyranose, 2,3,4,6-tetrabenzoate | 627466-84-6 | MFCD09841311 | 96.6% | 596.58 g/mol | C34H28O10 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW55687750MG
Encompass_Preferred
D-Galactopyranose, 2,3,4,6-tetrabenzoate is a benzoate-protected derivative of D-galactopyranose provided for glycobiology and carbohydrate chemistry applications. The tetrabenzoate protection increases hydrophobicity and handling stability, making the compound useful for organic synthesis, enzymatic assays, and analytical studies.
  • Used in glycobiology and carbohydrate chemistry research.
  • Benzoate protection improves solubility in organic media.
  • Molecular formula C34H28O10 and molecular weight 596.58 g/mol.
  • High purity (approximately 96.6%).
  • White to off-white solid; stable under recommended storage conditions.
  • Available in small gram-scale quantities for laboratory use.
Catalog No. 50-002-84900
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.