Learn More
Medchemexpress LLC D-Galactosamine pent 50g | 76375-60-5 | C16H23NO10 | 50 G
Supplier: Medchemexpress LLC HY11727150G
D-Galactosamine pentaacetate is an acetylated derivative of the amino sugar D-galactosamine used as a biochemical reagent and synthetic intermediate in carbohydrate and organic synthesis. The compound has chemical formula C16H23NO10 and a molecular weight of 389.35 g/mol.
- Acetyl-protected amino sugar suitable as a synthetic intermediate.
- Useful in glycosylation and derivatization reactions.
- Characterized by formula C16H23NO10 and molecular weight 389.35 g/mol.
- Intended for laboratory research and organic synthesis applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.