Learn More
Abcam D-(+)-Galactose, >98% Purity, 180.16 g/mol, 25 g
Description
- Molecular Weight: 180.16 g/mol
- Purity: >98%
- Description: C-4 epimer of glucose; aldohexose that serves as an energy source and as an essential component of glycolipids and glycoproteins
- Constituents: 100% D-(+)-Galactose
- CAS Number: 59-23-4
- Storage Conditions: Store at ambient
- Shipping Conditions: Ambient (can ship with ice)
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | Galactose |
| Shipping Condition | Ambient |
| Form | Solid |
| Content And Storage | Store at ambient temperature. |
| Buffer | 100% D-(+)-Galactose |
| Formulation | 100% D-(+)-Galactose |
| Quantity | 25 g |
| For Use With (Application) | Energy source, Glycolipid component, Glycoprotein component |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.