Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-(+)-Galactose, 98%

Catalog No. AAA128130B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA128130B 1000 g
AAA1281318 50 g
AAA1281330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA128130B Supplier Thermo Scientific Chemicals Supplier No. A128130B
Add to Cart

D-(+)-Galactose, CAS # 59-23-4, is a monosaccharide that is an important constituent of glycolipids and glycoproteins. It is also used as an energy source by organisms.

Galactose is a simple monosaccharide used as a component of the galactosyltransferase labeling buffer. It serves as an energy source and an important constituent of glycolipids and glycoproteins. Since it is a component of antigens, it plays a vital role in the determination of blood type within the ABO blood group system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • D-(+)-Galactose is a monosaccharide used as a component of the galactosyltransferase labeling buffer. The deficiency of galactosyl-1-phosphate uridyltransferase causes an error in galactose metabolism known as galactosemia, increasing the galactose levels in the blood
  • D-(+)-Galactose is a component of the disaccharide lactose and is released upon hydrolysis by β-galactosidase enzymes

Application

  • This compound can be used as a source of energy as it is an essential constituent of glycolipids and glycoproteins
  • It is a component of antigens which makes it important for determining blood type within the ABO blood group system
  • It has been used to induce the expression of uncoupling protein (UCP) in yeast transformants
  • It is frequently used as a source of carbon in vitro culture media
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-23-4
Molecular Formula C6H12O6
MDL Number MFCD00151230

Specifications

Melting Point 164°C to 168°C
Density 1.5
Quantity 1000 g
Beilstein 1724619
Merck Index 14,4335
Solubility Information Soluble in water.
Optical Rotation +80° (c=5 in Water, 24h)
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material D-(+)-Galactose
EINECSNumber 200-416-4
RTECSNumber LW5490000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.