Learn More
Medchemexpress LLC D-gluconic acid potassium | 299-27-4 | MFCD00064211 | 99.8% | 234.24 g/mol | C6H11KO7 | 100mg
SDP
Supplier:  Medchemexpress LLC HYY0569CR100MG
Encompass_Preferred
D-Gluconic acid (potassium) (Standard) is the analytical standard of D-Gluconic acid (potassium) This product is intended for research and analytical applications D-Gluconic acid potassium is an orally active carboxylic acid by the oxidation with antiseptic and chelating properties[1]
Catalog No. 50-002-69076