Learn More
Medchemexpress LLC Gluconic acid, monopotassium salt | 299-27-4 | MFCD00064211 | >99.0% | 234.25 g/mol | C6H11KO7 | 50mg
Supplier: Medchemexpress LLC HYY0569CR50MG
D-Gluconic acid (potassium) (Standard) is the analytical standard of D-Gluconic acid (potassium) This product is intended for research and analytical applications D-Gluconic acid potassium is an orally active carboxylic acid by the oxidation with antiseptic and chelating properties[1]
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.