Learn More
Medchemexpress LLC D-glucosamine 6-phosphate sodium | 70442-23-8 | MFCD00069826 | 99.9% | 281.13 g/mol | C6H13NNaO8P | 5mg
SDP
Supplier:  Medchemexpress LLC HYW7181365MG
Encompass_Preferred
D-Glucosamine 6-phosphate (sodium) (GlcN6P-Na) is a class of biochemical reagents used in glycobiology research Glycobiology studies the structure synthesis biology and evolution of sugars It involves carbohydrate chemistry enzymology of glycan formation and degradation protein-glycan recognition and the role of glycans in biological systems This field is closely related to basic research biomedicine and biotechnology[1]
Catalog No. 50-002-70393