Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Glucose 6-Phosphate Barium Salt Heptahydrate 98.0+%, TCI America™
SDP

Catalog No. G0052100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0052100MG 100 mg
G00521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0052100MG Supplier TCI America Supplier No. G0052100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58823-95-3
Molecular Formula C6H13O9P
MDL Number MFCD00221521

Specifications

Color White-Yellow
Quantity 100 mg
UN Number 1564
Formula Weight 395.45
Percent Purity ≥98.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material D-Glucose 6-Phosphate Barium Salt Heptahydrate
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.