Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-D-Glucose, MP Biomedicals

Catalog No. MP210095391 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210095391 1 kg
MP210095325 25 g
MP210095380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP210095391 Supplier MP Biomedicals, Inc Supplier No. 0210095391
Add to Cart
Add to Cart

  • β-D-Glucose is a main source of energy for living organisms
  • It is combined into glucosides, disaccharides, oligosaccharides, the polysaccharides (cellulose and starch), and glycogen

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-61-5
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00063989
InChI Key WQZGKKKJIJFFOK-VFUOTHLCSA-N
Synonym beta-d-glucose, beta-d-glucopyranose, glucoside, b-d-glucopyranose, beta-glucose, beta-dextrose, .beta.-d-glucopyranose, b-glucose, b-d-glucose, unii-j4r00m814d
PubChem CID 64689
ChEBI CHEBI:15903
IUPAC Name (2R,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES OC[C@H]1O[C@@H](O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 148°C to 155°C (literature)
Color White
Boiling Point ∼410.8°C (760 mmHg (Predicted) (literature))
Linear Formula C6H12O6
Quantity 1 kg
Solubility Information Soluble in water (1/20-clear solution) and alcohol (slightly).
Formula Weight 180.16
Percent Purity ≥80%
Physical Form Crystalline Powder
Chemical Name or Material β-D-GLUCOSE
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.