Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Glucurono-6,3-lactone 99.0+%, TCI America™
SDP

Catalog No. G0055100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0055100G 100 g
G005525G 25 g
G0055500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G0055100G Supplier TCI America Supplier No. G0055100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32449-92-6
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.124
MDL Number MFCD00135622
InChI Key UYUXSRADSPPKRZ-KKQCNMDGSA-N
Synonym d-glucuro-3,6-lactone
PubChem CID 91634076
IUPAC Name (2S)-2-[(2S,3S,4R)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde
SMILES C(=O)C(C1C(C(C(=O)O1)O)O)O

Specifications

Melting Point 178°C
Color White
pH 3.5 (10% H2O soln.)
Quantity 100 g
Formula Weight 176.12
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Glucurono-6,3-lactone
EINECSNumber (9)-0437
RTECSNumber LZ8930000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.