Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Isoascorbic acid, 100.01%, 98%, MP Biomedicals™

Catalog No. ICN10207390 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10207390 500 g
ICN10207380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10207390 Supplier MP Biomedicals, Inc Supplier No. 0210207390
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 89-65-6
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
MDL Number MFCD00005378
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
Synonym erythorbic acid, isoascorbic acid, d-araboascorbic acid, d-isoascorbic acid, araboascorbic acid, d-erythorbic acid, isovitamin c, neo-cebicure, saccharosonic acid, mercate 5
PubChem CID 54675810
ChEBI CHEBI:51438
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

Assay Percent Range 98%
Quantity 500 g
Formula Weight 176.1
Percent Purity 0.98
Grade Reagent
Physical Form Powder
Chemical Name or Material D-Isoascorbic acid
EINECSNumber 201-928-0
Recommended Storage Store at 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.