Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum nitrate nonahydrate, 99+%, for analysis

Catalog No. AC218285000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum

CAS: 7784-27-2 | AlN3O9·9H2O | 375.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-27-2
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
Synonym aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
PubChem CID 24567
IUPAC Name aluminum;trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]

Specifications

Melting Point 73.0°C
pH 2.5 to 3.5 (5% soln. at 20°C)
Physical Form Crystals
Quantity 500 g
Assay Percent Range 99% min. (Complexometry)
Molecular Formula AlN3O9·9H2O
Linear Formula Al(NO3)3·9H2O
Merck Index 15,347
Solubility Information Solubility in water: 64 g/100mL (25°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, practically insoluble in ethyl acetate and, pyridine
Molecular Weight (g/mol) 375.13
Formula Weight 375.13
Percent Purity ≥99%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Aluminum nitrate nonahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTRE or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.