Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Limonene, Food Grade, 95-97.5%, Spectrum™ Chemical
SDP

Catalog No. p-7157679 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
15 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-287 500 g Amber Glass Bottle
18-602-288 2.5 kg Amber Glass Bottle
18-602-289 15 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-287 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L1599500GM
Add to Cart
Add to Cart

L1599, 5989-27-5, C10H16

Spectrum™ Chemical D-Limonene, Food Grade , also known as 4-Isopropenyl-1-methylcyclohexene, is used as a flavorant and as a fragrance in perfumery, aftershave lotions, bath products and other such products. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 5989-27-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00062991
InChI Key XMGQYMWWDOXHJM-JTQLQIEISA-N
IUPAC Name (4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
SMILES CC(=C)[C@@H]1CCC(C)=CC1

Specifications

CAS Max % 100%
Boiling Point 175°C to 176°C
Melting Range -95.5°C to -74°C
Packaging Amber Glass Bottle
Refractive Index 1.471-1.474
Quantity 500 g
UN Number UN2052
Optical Rotation +96.0° to +104°
Percent Purity 95 to 97.5%
Grade Food Grade
Physical Form Liquid
Chemical Name or Material D-Limonene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.