CAS RN 5989-27-5
(R)-(+)-Limonene, 96%
CAS: 5989-27-5 Molecular Formula: C10H16 Molecular Weight (g/mol): 136.24 MDL Number: MFCD00062991 InChI Key: XMGQYMWWDOXHJM-JTQLQIEISA-N Synonym: d-limonene,+-limonene,r-+-limonene,+-4r-limonene,+-carvene,+-dipentene,citrene,+-p-mentha-1,8-diene,4r-limonene,d-+-limonene PubChem CID: 440917 ChEBI: CHEBI:15382 IUPAC Name: (4R)-1-methyl-4-prop-1-en-2-ylcyclohexene SMILES: CC(=C)[C@@H]1CCC(C)=CC1
D-Limonene, 96.9%, MP Biomedicals
CAS: 5989-27-5 Molecular Formula: C10H16 Molecular Weight (g/mol): 136.24 MDL Number: MFCD00062991 InChI Key: XMGQYMWWDOXHJM-JTQLQIEISA-N Synonym: d-limonene,+-limonene,r-+-limonene,+-4r-limonene,+-carvene,+-dipentene,citrene,+-p-mentha-1,8-diene,4r-limonene,d-+-limonene PubChem CID: 440917 ChEBI: CHEBI:15382 SMILES: CC(=C)[C@@H]1CCC(C)=CC1
(+)-Limonene, 96%, unstabilized
CAS: 5989-27-5 Molecular Formula: C10H16 Molecular Weight (g/mol): 136.24 MDL Number: MFCD00062991 InChI Key: XMGQYMWWDOXHJM-JTQLQIEISA-N Synonym: d-limonene,+-limonene,r-+-limonene,+-4r-limonene,+-carvene,+-dipentene,citrene,+-p-mentha-1,8-diene,4r-limonene,d-+-limonene PubChem CID: 440917 ChEBI: CHEBI:15382 SMILES: CC(=C)[C@@H]1CCC(C)=CC1
(+)-Limonene 99.0+%, TCI America™
CAS: 5989-27-5 Molecular Formula: C10H16 Molecular Weight (g/mol): 136.24 MDL Number: MFCD00062991 InChI Key: XMGQYMWWDOXHJM-JTQLQIEISA-N Synonym: d-limonene,+-limonene,r-+-limonene,+-4r-limonene,+-carvene,+-dipentene,citrene,+-p-mentha-1,8-diene,4r-limonene,d-+-limonene PubChem CID: 440917 ChEBI: CHEBI:15382 IUPAC Name: (4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene SMILES: CC(=C)[C@@H]1CCC(C)=CC1
D-Limonene, Food Grade, 95-97.5%, Spectrum™ Chemical
CAS: 5989-27-5 Molecular Formula: C10H16 Molecular Weight (g/mol): 136.24 MDL Number: MFCD00062991 InChI Key: XMGQYMWWDOXHJM-JTQLQIEISA-N IUPAC Name: (4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene SMILES: CC(=C)[C@@H]1CCC(C)=CC1
(R)-(+)-Limonene, ≥99% (sum of enantiomers, GC), MilliporeSigma™ Supelco™
(R)-(+)-Limonene is a terpenoid compound that is abundantly present in many plants such as rosemary. It is highly associated with plant and insect interactions due to its tendency to attract insects.
(+)-Limonene 95.0+%, TCI America™
CAS: 5989-27-5 Molecular Formula: C10H16 Molecular Weight (g/mol): 136.24 MDL Number: MFCD00062991 InChI Key: XMGQYMWWDOXHJM-JTQLQIEISA-N Synonym: d-limonene,+-limonene,r-+-limonene,+-4r-limonene,+-carvene,+-dipentene,citrene,+-p-mentha-1,8-diene,4r-limonene,d-+-limonene PubChem CID: 440917 ChEBI: CHEBI:15382 IUPAC Name: (4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene SMILES: CC(=C)[C@@H]1CCC(C)=CC1
Sigma Aldrich ™-(+)-Limonene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
PROTOCOL™ Histosolv-X, Fisher Healthcare
CAS: 5989-27-5 Molecular Formula: C10H16 Molecular Weight (g/mol): 136.24 MDL Number: MFCD00062991 InChI Key: XMGQYMWWDOXHJM-JTQLQIEISA-N Synonym: d-limonene,+-limonene,r-+-limonene,+-4r-limonene,+-carvene,+-dipentene,citrene,+-p-mentha-1,8-diene,4r-limonene,d-+-limonene PubChem CID: 440917 ChEBI: CHEBI:15382 IUPAC Name: (4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene SMILES: CC(=C)[C@@H]1CCC(C)=CC1
Ricca Chemical Company D-Limonene, Natural Isolate, Ricca Chemical
D-Limonene, Natural Isolate is a natural isolate food additive.
Decon™ CitriSolv™ Hybrid Solvent and Clearing Agent
Can be used as an alternative to xylene and ethyl acetate. CitriSolv Hybrid is soluble with absolute alcohol and will dissolve paraffin, glue, adhesives and mounting media.