Learn More
Selleck Chemical LLC CP 640186
SDP
Supplier:  Selleck Chemical LLC S67535MG
Encompass_Preferred
CP 640186 is an isozyme-nonselective ACC (Acetyl-CoA carboxylase) inhibitor with IC50 values of 53 and 61 nM for rat liver ACC1 and rat skeletal muscle ACC2 respectively
Catalog No. 50-000-01880
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.