Learn More
Medchemexpress LLC Benzeneacetic acid, α-hydroxy-, (αR)- | 611-71-2 | MFCD00064251 | 99.9% | 152.15 g/mol | C8H8O3 | 25g
SDP
Supplier:  Medchemexpress LLC HYY058525G
Encompass_Preferred
D-(-)-Mandelic acid is an orally active alpha hydroxycarboxylic acid that can be isolated from bitter almonds and Indian chestnut trees It has antioxidant and antibacterial properties and is expected to play an important role in the treatment of rheumatoid arthritis[1][4] /br
Catalog No. 50-002-68889