Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Mannitol, ≥99.0% (sum of enantiomers, HPLC); ≥99.9999% (metals basis), MilliporeSigma™ Supelco™

Catalog No. 111015264 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5264 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5264 Supplier MilliporeSigma Supelco Supplier No. 7851350G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ultra-pure sugar alcohol for boron determination and analytical reference standard applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
Synonym Mannite

Specifications

Melting Point 166°C to 169°C; 167°C to 170°C
CAS 69-65-8
Linear Formula C6H14O6
Quantity 50 g
Formula Weight 182.17
Percent Purity ≥99.0% (sum of enantiomers, HPLC); ≥99.9999% (metals basis)
Grade Not Available
Physical Form Powder
RTECSNumber OP2060000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.