Learn More
AdipoGen D Mannose
SDP
Supplier:  AdipoGen CDXM0020G100
Encompass_Preferred
Chemical. CAS 3458-28-4. Formula C6H12O6. MW 180.16. Synthetic. Mannose is an aldohexose carbohydrate. This product may be used to study the effects of exogenous mannose on in vitro processes such as glycosylation.
Catalog No. 50-256-0562
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.