Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-NMAPPD ((1R,2R)-B13) | 35922-06-6 | MFCD08062172 | 99.7% | 422.56 | C23H38N2O5 | 10MG
SDP

Supplier:  Medchemexpress LLC HY12496210MG

Encompass_Preferred

D-NMAPPD ((1R,2R)-B13) is a research-grade small molecule inhibitor of acid ceramidase. It modulates NMDA receptor properties by enhancing endogenous ceramide production and has demonstrated anticancer activity in cellular studies. For research use only; not for human or veterinary use.

  • Acid ceramidase inhibitor that induces apoptosis in cell models.
  • Modulates NMDA receptor function via increased ceramide production.
  • CAS number: 35922-06-6.
  • Molecular formula: C23H38N2O5; molecular weight: 422.56.
  • High purity suitable for research: 99.7%.
  • Available in solid and solution formats, including 10 MG and 10 mM in DMSO solutions.

Catalog No. 50-002-37311


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.