Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Pantethine, 98%, Spectrum™ Chemical
SDP

Catalog No. 18602339 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-339 1 g Amber Glass Bottle
18-602-340 5 g Amber Glass Bottle
18-602-341 25 g Amber Glass Bottle
18-602-342 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-602-339 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P10241GM
Add to Cart
Add to Cart

P1024, 16816-67-4, C22H42N4O8S2

Spectrum™ Chemical D-Pantethine , also known as Bis-pantethine, can be used to treat acne and improve the blood-cholesterol profile. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 16816-67-4
Molecular Formula C22H42N4O8S2
Molecular Weight (g/mol) 554.72
InChI Key DJWYOLJPSHDSAL-ROUUACIJSA-N
IUPAC Name (2R)-N-[2-({2-[(2-{3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanamido}ethyl)disulfanyl]ethyl}carbamoyl)ethyl]-2,4-dihydroxy-3,3-dimethylbutanamide
SMILES CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)NCCSSCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)CO

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 1 g
Percent Purity 98%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.