Learn More
D-Phenylalanine tert-butyl ester hydrochloride, 98%
Description
D-Phenylalanine tert-butyl ester hydrochloride is used as a fine chemical intermediate, agrochemical &pharmceutical intermediate. It has anti - static and anti- corrosion properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
D-Phenylalanine tert-butyl ester hydrochloride is used as a fine chemical intermediate, agrochemical &pharmceutical intermediate. It has anti - static and anti- corrosion properties.
Solubility
Soluble in water.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Specifications
| Solubility Information | Soluble in water. |
| Quantity | 250 mg |
| Formula Weight | 257.76 |
| Percent Purity | 98% |
| Physical Form | Solid |
| Chemical Name or Material | D-Phenylalanine tert-butyl ester hydrochloride |
Safety and Handling
| TSCA | No |
| storageNote1 | -30° to -10°C |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.