Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Phenylalaninol, 98%, Thermo Scientific Chemicals

Catalog No. aal09697md
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL09697MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL09697MD Supplier Thermo Scientific Chemicals Supplier No. L09697MD

D-Phenylalaninol is used as a chiral auxiliary for asymmetric Michael reactions. It is an enantiomer of L-Phenylalaninol, an inhibitor of intestinal Phenylalanine absorption. It acts as an inhibiting agent to the enzymes which are responsible for the breakdown of endorphins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Phenylalaninol is used as a chiral auxiliary for asymmetric Michael reactions. It is an enantiomer of L-Phenylalaninol, an inhibitor of intestinal Phenylalanine absorption. It acts as an inhibiting agent to the enzymes which are responsible for the breakdown of endorphins.

Solubility
Soluble in Dichloromethane, Ethyl Acetate, Methanol.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with carbondioxide, air, acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5267-64-1
Molecular Formula C9H13NO
MDL Number MFCD00064298
Synonym d-phenylalaninol, r-2-amino-3-phenylpropan-1-ol, d-penylalaninol, r-+-2-amino-3-phenyl-1-propanol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, +-d-phenylalaninol, r-2-amino-3-phenyl-1-propanol, d-+-phenylalaninol, r-phenylalaninol

Specifications

Melting Point 89°C to 94°C
Quantity 250 mg
Optical Rotation +23 ± 2° (c=5 in Ethanol)
UN Number UN3259
Beilstein 4665408
Formula Weight 151.21
Percent Purity 98%
Chemical Name or Material D-Phenylalaninol
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.