Learn More
Thermo Scientific Chemicals D-Pinitol, 95%
Description
It is a precursor to biologically active fluorinated isosteres of inositol, which show cell growth inhibitory properties and has shown antidiabetic properties in mice. It is believed to be a salt stress regulator in a wide range of plants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is a precursor to biologically active fluorinated isosteres of inositol, which show cell growth inhibitory properties and has shown antidiabetic properties in mice. It is believed to be a salt stress regulator in a wide range of plants.
Solubility
Soluble in water.
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications
Specifications
| Melting Point | 178°C to 185°C |
| Quantity | 1 g |
| Merck Index | 14,10361 |
| Solubility Information | Soluble in water. |
| Optical Rotation | +60° (c=1 in Water) |
| Formula Weight | 194.18 |
| Percent Purity | 95% |
| Chemical Name or Material | D-Pinitol |
Safety and Handling
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.