Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Proline Benzyl Ester Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. P17261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17261G 1 g
P17265G 5 g
P172625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P17261G Supplier TCI America Supplier No. P17261G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53843-90-6
Molecular Formula C12H16ClNO2
Molecular Weight (g/mol) 241.715
MDL Number MFCD00153458
InChI Key NEDMOHHWRPHBAL-RFVHGSKJSA-N
Synonym h-d-pro-obzl hcl, h-d-pro-obzl.hcl, d-proline benzyl ester hydrochloride, d-proline, phenylmethyl ester, hydrochloride, r-benzyl pyrrolidine-2-carboxylate hydrochloride, benzyl 2r-pyrrolidine-2-carboxylate hydrochloride, h-d-pro-obzl-hcl, h-d-pro-obzl inverted exclamation mark currencyhcl, d-proline benzylester hydrochloride, d-proline-benzyl-ester hydrochloride
PubChem CID 22827650
IUPAC Name benzyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
SMILES C1CC(NC1)C(=O)OCC2=CC=CC=C2.Cl

Specifications

Color White
Quantity 1 g
Formula Weight 241.72
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material D-Proline Benzyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.