Learn More
Sigma Organic Chemistry D-(-)-Ribose | 25G | 50-69-1 | MFCD00135453
Supplier: Sigma Organic Chemistry R175725GA

D-(-)-Ribose, 25G
About This Item:
Linear Formula: C5H10O5
Storage: room temp
EINECS Number: 200-059-4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.