Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-Ribose-5-phosphate disodium salt hydrate, 85%

Catalog No. AAJ6681503
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6681503 1 g
AAJ66815MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6681503 Supplier Thermo Scientific Chemicals Supplier No. J6681503
Add to Cart

CAS: 18265-46-8 | C5H9Na2O8P | 274.07 g/mol

Ribose-5-phosphate is an intermediate of the oxidative branch of the Pentose Phosphate Pathway. Ribose-5-phosphate is used in the synthesis of nucleotides and nucleic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ribose-5-phosphate is an intermediate of the oxidative branch of the Pentose Phosphate Pathway. Ribose-5-phosphate is used in the synthesis of nucleotides and nucleic acids.

Solubility
Soluble in water at 50mg/ml

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18265-46-8
Molecular Formula C5H9Na2O8P
Molecular Weight (g/mol) 274.07
MDL Number MFCD00135931
InChI Key RHCIGTFSCDIJGH-CHEKPSGWSA-L
Synonym d-ribofuranose5-phosphatedisodiumsalt
PubChem CID 131875268
IUPAC Name sodium;[(2R,3R,4R)-2,3,4-trihydroxy-5-oxopentyl] dihydrogen phosphate
SMILES [Na+].[Na+].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O

Specifications

Color White to Yellow
Quantity 1 g
Solubility Information Soluble in water at 50mg/ml
Formula Weight 310.1
Percent Purity 85%
Physical Form Powder
Chemical Name or Material D-Ribose-5-phosphate disodium salt hydrate
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.