Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ D-(-)-Ribose, Calbiochem™,

Catalog No. 7410100GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-101-00GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 74-101-00GM Supplier MilliporeSigma™ Supplier No. 7410100GM

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50-69-1
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00135453
InChI Key HMFHBZSHGGEWLO-ZHZWXSSZNA-N
Synonym 2r,3r,4r-2,3,4,5-tetrahydroxypentanal, aldehydo-d-ribose, ribose, d, d--ribose, aldehydo-d-ribo-pentose, ribo-2,3,4,5-tetrahydroxyvaleraldehyde, d, d-ribo-2,3,4,5-tetrahydroxyvaleraldehyde, ribose 9ci, dsstox_cid_23917, dsstox_gsid_43917
PubChem CID 5311110
ChEBI CHEBI:47014
IUPAC Name (3R,4S,5R)-5-(hydroxymethyl)oxolane-2,3,4-triol
SMILES OC[C@H]1OC(O)[C@H](O)[C@@H]1O

Specifications

Color White
Quantity 100 g
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material D-(-)-Ribose
Grade Molecular biology grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.