Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-ribulose, 99.7%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN15202890 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
500 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15202890 500 mg
ICN15202810 10 mg
ICN15202825 25 mg
ICN15202880 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15202890 Supplier MP Biomedicals, Inc Supplier No. 0215202890
Add to Cart
Add to Cart

Chemical Identifiers

CAS 488-84-6
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key ZAQJHHRNXZUBTE-NQXXGFSBSA-N
Synonym d-ribulose, ribulose, d-erythro-pent-2-ulose, d-erythropentulose, d-erythro-pentulose, unii-z7u4kg0138, 3r,4r-1,3,4,5-tetrahydroxypentan-2-one, arabinulose, araboketose, erythropentulose
PubChem CID 151261
ChEBI CHEBI:17173
IUPAC Name (3R,4R)-1,3,4,5-tetrahydroxypentan-2-one
SMILES C(C(C(C(=O)CO)O)O)O

Specifications

Color Colorless
Quantity 500 mg
Formula Weight 150.1
Percent Purity 0.997
Physical Form Liquid
Chemical Name or Material D-ribulose, For HPLC analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.