Learn More
Cole-Parmer Digi-Sense Armored Liquid-In-Glass Thermometer, -40 to 120°F, 76 mm Immersion; Organic Liquid Fill
Supplier: Cole-Parmer 0807794
Ideal for use in application where thermometers needs added protection from minor shocks. Nickel/plated brass armor shell protects these thermometers from accidental breakage during use and storage. The armor case has a screw cap with ring top and viewing window; the lower section that surrounds the bulb has small openings for circulation. Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include statement of accuracy and a triangular-style antiroll fitting.