Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(-)-Salicin, 99%

Catalog No. aaa1652406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1652406 5 g
AAA1652414 25 g
AAA1652422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1652406 Supplier Thermo Scientific Chemicals Supplier No. A1652406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 138-52-3 | C13H18O7 | 286.28 g/mol

Salicin is used as an analgesic and antipyretic. It is also used as a substrate for beta-glucosidase and an inhibitor of Cox. It finds application as an anti-inflammatory agent. It is involved in antipyretic action without causing gastric injury.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Salicin is used as an analgesic and antipyretic. It is also used as a substrate for beta-glucosidase and an inhibitor of Cox. It finds application as an anti-inflammatory agent. It is involved in antipyretic action without causing gastric injury.

Solubility
Soluble in water, alkalies, pyridine, glacial acetic acid and hot ethanol. Insoluble in ether and chloroform.

Notes
Store in cool place. Light and heat sensitive. Air senstive and moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-52-3
Molecular Formula C13H18O7
Molecular Weight (g/mol) 286.28
MDL Number MFCD00006590
InChI Key NGFMICBWJRZIBI-UJPOAAIJSA-N
Synonym salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside
PubChem CID 439503
ChEBI CHEBI:17814
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
SMILES C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 200°C to 203°C
Odor Odorless
Assay Percent Range 99%
Quantity 5 g
Beilstein 89593
Merck Index 14,8324
Solubility Information Soluble in water,alkalies,pyridine,glacial acetic acid and hot ethanol. Insoluble in ether and chloroform.
Optical Rotation −62.5° (c=3 in Water)
Formula Weight 286.28
Percent Purity 99%
Chemical Name or Material D-(-)-Salicin
Show More Show Less
Hazard Category H317
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 205-331-6
RTECSNumber LZ5901700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.