Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Serine, 98%

Catalog No. AC227070050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227070050 5 g Glass Bottle
AC227070250 25 g Glass Bottle
AC227071000 100 g Glass Bottle
AC227075000 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC227070050 Supplier Thermo Scientific Chemicals Supplier No. 227070050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 312-84-5 | C3H7NO3 | 105.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312-84-5
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00004269
InChI Key MTCFGRXMJLQNBG-UWTATZPHSA-N
Synonym d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin
PubChem CID 71077
ChEBI CHEBI:16523
IUPAC Name (2R)-2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point 220.0°C
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula HOCH2CH(NH2)CO2H
Specific Rotation − 14.75
Solubility Information Solubility in water: soluble in water: 346g/L (20°C). Other solubilities: insoluble in alcohol, ether and benzene., soluble in mineral acids., insoluble in organic solvents.
Specific Rotation Condition −14.75° (20°C c=10 2 N HCl)
Quantity 5 g
Formula Weight 105.09
Percent Purity 98%
Chemical Name or Material D-Serine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.