CAS RN 312-84-5
D-Serine, 98%
CAS: 312-84-5 Molecular Formula: C3H7NO3 Molecular Weight (g/mol): 105.093 MDL Number: MFCD00004269 InChI Key: MTCFGRXMJLQNBG-UWTATZPHSA-N Synonym: d-serine,r-2-amino-3-hydroxypropanoic acid,2r-2-amino-3-hydroxypropanoic acid,serine d-form,r-serine,serine, d,d-+-serine,h-d-ser-oh,d-2-amino-3-hydroxypropionic acid,d-serin PubChem CID: 71077 ChEBI: CHEBI:16523 IUPAC Name: (2R)-2-amino-3-hydroxypropanoic acid SMILES: C(C(C(=O)O)N)O
Thermo Scientific Chemicals D-Serine, 99%
CAS: 312-84-5 Molecular Formula: C3H7NO3 Molecular Weight (g/mol): 105.093 MDL Number: MFCD00004269 InChI Key: MTCFGRXMJLQNBG-UWTATZPHSA-N Synonym: d-serine,r-2-amino-3-hydroxypropanoic acid,2r-2-amino-3-hydroxypropanoic acid,serine d-form,r-serine,serine, d,d-+-serine,h-d-ser-oh,d-2-amino-3-hydroxypropionic acid,d-serin PubChem CID: 71077 ChEBI: CHEBI:16523 IUPAC Name: (2R)-2-amino-3-hydroxypropanoic acid SMILES: C(C(C(=O)O)N)O
D-Serine 98.5+%, TCI America™
CAS: 312-84-5 Molecular Formula: C3H7NO3 Molecular Weight (g/mol): 105.093 MDL Number: MFCD00004269 InChI Key: MTCFGRXMJLQNBG-UWTATZPHSA-N Synonym: d-serine,r-2-amino-3-hydroxypropanoic acid,2r-2-amino-3-hydroxypropanoic acid,serine d-form,r-serine,serine, d,d-+-serine,h-d-ser-oh,d-2-amino-3-hydroxypropionic acid,d-serin PubChem CID: 71077 ChEBI: CHEBI:16523 IUPAC Name: (2R)-2-amino-3-hydroxypropanoic acid SMILES: C(C(C(=O)O)N)O
R&D Systems™ D-Serine
CAS: 312-84-5 Molecular Formula: C3H7NO3