Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Sorbitol, Anhydrous, Cell Culture Reagent, ≥98%, MP Biomedicals™

Catalog No. ICN19474205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19474205 5 kg
02194742-5 500 g
ICN19474201 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19474205 Supplier MP Biomedicals, Inc Supplier No. 0219474205
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Sorbitol, Anhydrous, Cell Culture Reagent, ≥98% is used for inducing osmotic stress.

  • Used for washing spheroplasts and in isoelectric focusing to minimize endoosmotic flow in agarose gels.
  • May be used to induce osmotic stress.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, glucarine, diakarmon, multitol, sorbilande
PubChem CID 5780
ChEBI CHEBI:17924

Specifications

Melting Point 230°F
Density 1.49 g/cm3 at 59°F
pH 7.0
Boiling Point Very High
Flash Point 542°F
Quantity 5 kg
Solubility Information Water Solubility: 2,750,000 mg/L at 30°C
Optical Rotation −4 to 0 ° (c=2, water)
Vapor Pressure 9.9X10-9 mm Hg at 25°C
Percent Purity ≥98%
Grade Cell Culture Grade
Physical Form Powder
Conductivity <20.0 μS/cm
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.