Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-Tagatose, 99%

Catalog No. AAB2119201
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2119201 0.25 g
AAB2119203 1 g
AAB2119206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2119201 Supplier Thermo Scientific Chemicals Supplier No. B2119201
Add to Cart

CAS: 87-81-0 | C6H12O6 | 180.156 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-81-0
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00134449
InChI Key BJHIKXHVCXFQLS-PQLUHFTBSA-N
Synonym 3s,4s,5r-1,3,4,5,6-pentahydroxyhexan-2-one, lyxo-2-hexulose, tagatose, d, d--tagatose, keto-d-tagatose, tagatose nf, unii-t7a20y888y, d-lyxo-hex-2-ulose, naturlose
PubChem CID 92092
ChEBI CHEBI:47693
IUPAC Name (3S,4S,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Melting Point 129°C to 135°C
Quantity 0.25 g
Beilstein 1724555
Merck Index 14,9030
Optical Rotation −4.7° (c=1 in Water)
Formula Weight 180.16
Percent Purity 99%
Chemical Name or Material D-Tagatose
EINECSNumber 201-772-3
TSCA No

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.