Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Talose, 99%, Thermo Scientific™

Catalog No. AC257882500
Change view
Click to view available options
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC257882500 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC257882500 Supplier Thermo Scientific Chemicals Supplier No. 257882500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2595-98-4
Assay Percent Range 99%
Molecular Formula C6H12O6
Quantity 250mg
MDL Number MFCD00135834
Specific Rotation Condition +20° (20°C c=1,water)
Synonym d-+-talose, aldehydo-d-talose, 2s,3s,4s,5r-2,3,4,5,6-pentahydroxyhexanal
InChI Key GZCGUPFRVQAUEE-UHFFFAOYNA-N
SMILES OCC(O)C(O)C(O)C(O)C=O
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal
Molecular Weight (g/mol) 180.16
PubChem CID 99459
ChEBI CHEBI:68461
Formula Weight 180.16
Chemical Name or Material D(+)-Talose, 99%
Show More Show Less
EINECSNumber 219-996-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.