Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Trehalose dihydrate, >99%, MP Biomedicals™

Catalog No. ICN19475610 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19475610 10 g
ICN19475605 5 g
ICN19475625 25 g
ICN19475680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19475610 Supplier MP Biomedicals, Inc Supplier No. 0219475610
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6138-23-4
Molecular Formula C12H26O13
Molecular Weight (g/mol) 378.33
MDL Number MFCD00071594
InChI Key DPVHGFAJLZWDOC-DJCWUSJTNA-N
Synonym trehalose dihydrate, d-+-trehalose dihydrate, a,a-trehalose, d +-trehalose dihydrate, unii-7yin7j07x4, alpha,alpha-trehalose dihydrate, d-trehalose dihydrate, mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside, alp,alp.-trehalose, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triol
PubChem CID 181978
SMILES O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Color White
Assay Percent Range >99%
Quantity 10 g
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
Formula Weight 378.3
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Trehalose dihydrate
EINECSNumber 202-739-6
Recommended Storage Store at Room Temperature (15 to 30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.